Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-1/CD54 Rabbit anti-Rat, Janelia Fluor 646, Clone: 112, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290847JF646
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Rat samples. It is validated for Sandwich ELISA Detection.Specifications
ICAM-1/CD54 | |
Monoclonal | |
Janelia Fluor 646 | |
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ICAM-1/CD54 (Q00238-1; Met1-Thr493). | |
0.1 mL | |
Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers | |
3383 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
112 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title