Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2/CD102 Rabbit anti-Human, DyLight 755, Clone: 15, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289421IR
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
ICAM-2/CD102 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
15 | |
Sandwich ELISA Detection | |
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (NP_000864.2; Met1-Gln223). | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3384 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title