Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-2/CD102 Rabbit anti-Human, FITC, Clone: 41, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289424F

Catalog No. NB066825


Add to cart

Description

Description

ICAM-2/CD102 Monoclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for ELISA
Specifications

Specifications

ICAM-2/CD102
41
ELISA
Purified
Rabbit
IgG
0.1 mL
Adaptive Immunity, Immunology
Primary
3384
ELISA
FITC
PBS
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Recombinant Human ICAM-2/CD102 protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit