Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2 Recombinant Rabbit Monoclonal Antibody (033), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$221.00 - $430.00
Specifications
Antigen | ICAM-2 |
---|---|
Clone | 033 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543107
![]() |
Thermo Scientific
MA543107 |
100 μL |
Each for $430.00
|
|
PIMA543444
![]() |
Thermo Scientific
MA543444 |
50 μL |
Each for $221.00
|
|
Description
Specificity: Human ICAM-2/CD102
ICAM-2 Recombinant Monoclonal antibody specifically detects ICAM-2 in Human samples. It is validated for Western BlotSpecifications
ICAM-2 | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 | |
ICAM2 | |
IgG | |
Protein A | |
Antibody |
033 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P13598 | |
3384 | |
Recombinant Human ICAM-2/CD102 protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
ICAM2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title