Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ICAM-3/CD50 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795715PLATES

 View more versions of this product

Catalog No. NB010182


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ICAM-3/CD50.
Specifications

Specifications

HRP
Sandwich ELISA
62.5 pg/mL
3385
RUO
ICAM-3/CD50
Cellular Markers, Immunology
Human
ICAM3
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.
Antibody Pairs
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ICAM-3/CD50.
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL mouse anti-ICAM-3/CD50 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.5 mg/mL of mouse anti-ICAM-3/CD50 monoclonal antibody.
  • Standard: Each vial contains 40 ng of recombinant ICAM-3/CD50.
  • Storage is content dependent.
    Mouse
    5 Plates
    62.5-4000 pg/mL
    CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit