Learn More
ICAM-3/CD50 Mouse anti-Human, Alexa Fluor 405, Clone: MM0334-8V23, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212170AF405
Catalog No. NB212170405
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
ICAM-3/CD50 Monoclonal antibody specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot.
Specifications
Specifications
ICAM-3/CD50
MM0334-8V23
Western Blot
Purified
ICAM3
Human ICAM3
Protein G purified
Cellular Markers, Immunology
Primary
3385
Human
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody was selected for its ability to detect human CD50.