Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
ICAM-3/CD50 Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0334-8V23, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212170AF750
Catalog No. NP212170A70
Description
Description
ICAM-3/CD50 Monoclonal antibody specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot.
Specifications
Specifications
ICAM-3/CD50
MM0334-8V23
Western Blot
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody was selected for its ability to detect human CD50.
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
ICAM3
Human ICAM3
0.1 mL
Primary
3385
Human