Learn More
ICAM-3/CD50 Mouse anti-Human, Janelia Fluor 549, Clone: By44, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250432JF549
Catalog No. NBP50432549
Add to cart
Description
Description
ICAM-3/CD50 Monoclonal antibody specifically detects ICAM-3/CD50 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
ICAM-3/CD50
By44
Flow Cytometry, Immunohistochemistry
Purified
ICAM3
B cell lymphoma cells
Protein G purified
Cellular Markers, Immunology
Primary
3385
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Human