Learn More
ICAM-4 Sheep anti-Human, PE, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems FAB7179P025
 View more versions of this product
Catalog No. FAB7179P025
Add to cart
Description
Description
ICAM-4 (Intercellular Adhesion Molecule 4), also known as Landsteiner-Wiener glycoprotein and CD242, is a 42kDa member of the ICAM family, Ig superfamily of proteins. It is expressed on erythrocytes and erythroblasts, and serves as a receptor for LFA-1, Mac-1, and CD11c/CD18, plus α4β1 and alpha-V containing integrins. ICAM-4 is suggested to bind to Mac-1 on macrophages, allowing for its phagocytosis in senescence. Mature human ICAM-4 is a 249 amino acid (aa) type I transmembrane glycoprotein. It possesses a 218 aa extracellular region (aa 23-240) that contains two C2-type Ig-like domains (aa 62-124 and 146-217), and a 10 aa C-terminal cytoplasmic tail. ICAM-4 may form 85kDa homodimers. There are three potential isoform variants. One shows a five aa substitution for aa 233-271, a second contains a 15 aa substitution for aa 14-29, and a third possesses a 141 aa substitution for aa 132-271. Over aa 31-240, human ICAM-4 shares 71% aa sequence identity with mouse ICAM-4.
Specifications
Specifications
ICAM-4
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
ICAM4
Chinese hamster ovary cell line CHO-derived recombinant human ICAM-4, Ala31-Ala240, Accession # Q14773
Antigen Affinity-purified
RUO
Primary
3386
Human
Flow Cytometry
Flow Cytometry 10 uL/10^6 cells
Q14773
CD242, CD242 antigen, ICAM4, ICAM-4, intercellular adhesion molecule 4, intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), intercellular adhesion molecule 4 (LW blood group), intercellular adhesion molecule 4, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group antigen a, Landsteiner-Wiener blood group glycoprotein, LWLW blood group protein
Sheep
IgG
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Polyclonal
Detects human ICAM-4 in direct ELISAs and Western blots.