Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICAM-4 PE-conjugated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Supplier: R&D Systems FAB7179P100
This item is not returnable.
View return policy
Description
ICAM-4 Polyclonal specifically detects ICAM-4 in Human samples. It is validated for Flow Cytometry.Specifications
ICAM-4 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
Q14773 | |
ICAM4 | |
Chinese hamster ovary cell line CHO-derived recombinant human ICAM-4 Ala31-Ala240 Accession # Q14773 | |
100 Tests | |
Primary | |
Detects human ICAM-4 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD242, CD242 antigen, ICAM4, ICAM-4, intercellular adhesion molecule 4, intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), intercellular adhesion molecule 4 (LW blood group), intercellular adhesion molecule 4, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group antigen a, Landsteiner-Wiener blood group glycoprotein, LWLW blood group protein | |
Sheep | |
Affinity Purified | |
RUO | |
3386 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction