Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-4 Sheep anti-Human, PE, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

$177.50 - $345.00

Specifications

Antigen ICAM-4
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Format Affinity Purified
Applications Flow Cytometry
Conjugate R-PE
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB7179P025
View Documents
R&D Systems
FAB7179P025
25 Tests
Each for $177.50
Add to cart
 
FAB7179P100
View Documents
R&D Systems
FAB7179P100
100 Tests
Each for $345.00
Add to cart
 
Description

Description

ICAM-4 Polyclonal specifically detects ICAM-4 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ICAM-4
Affinity Purified
R-PE
IgG
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Flow Cytometry
Sheep
Antigen Affinity-purified
RUO
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit