Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-5 Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0250-3J37, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212361AF350

Catalog No. NP212361A30


Add to cart

Description

Description

ICAM-5 Monoclonal antibody specifically detects ICAM-5 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

ICAM-5
RM0250-3J37
Western Blot
Conjugated, Purified
ICAM5
Mouse ICAM5
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin
Rat
IgG2a
0.1 mL
Primary
7087.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit