Learn More
ICAM-5 Rat anti-Mouse, Alexa Fluor 647, Clone: RM0250-3J37, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212361AF647
Catalog No. NB212361647
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
ICAM-5 Monoclonal antibody specifically detects ICAM-5 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
ICAM-5
RM0250-3J37
Western Blot
Purified
ICAM5
Mouse ICAM5
Protein G purified
Store at 4C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin
Rat
IgG2a
0.1 ml
Primary
7087