Learn More
ICAM-5 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0250-3J37, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212361AF750
Catalog No. NP212361A70
Add to cart
Description
Description
ICAM-5 Monoclonal antibody specifically detects ICAM-5 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
ICAM-5
RM0250-3J37
Western Blot
ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin
Rat
IgG2a
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
ICAM5
Mouse ICAM5
0.1 mL
Primary
7087