Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-5 Rat anti-Mouse, Clone: RM0250-3J37, Novus Biologicals™
Rat Monoclonal Antibody
Specifications
Antigen | ICAM-5 |
---|---|
Clone | RM0250-3J37 |
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP212361
|
Novus Biologicals
NBP212361 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB019192
|
Novus Biologicals
NBP2123610.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
ICAM-5 Monoclonal antibody specifically detects ICAM-5 in Mouse samples. It is validated for Western Blot.Specifications
ICAM-5 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin | |
ICAM5 | |
IgG2a | |
Protein G purified |
RM0250-3J37 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Q60625 | |
7087 | |
Mouse ICAM5 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title