Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-1 Mouse anti-Human, FITC, Clone: 1H4, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15748

Catalog No. 50-113-7090


Add to cart

Description

Description

ICAM-1 (intercellular adhesion molecule) has 7 potential N-linked glycosylation sites. It is a single chain glycoprotein of Ig supergene family, present on unstimulated endo-thelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1).
Specifications

Specifications

ICAM-1
Monoclonal
FITC
ICAM1
Liquid
ICAM1
Human CD54
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
1H4
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P05362
BB2, P3.58, ICAM-1, cell surface glycoprotein P3.58, intercellular adhesion molecule 1, human rhinovirus receptor, major group rhinovirus receptor
Mouse
IgG2b
100 tests
4° C
Primary
3383
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit