Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM3 Mouse anti-Human, Unconjugated, Clone: 186-2G9, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB21240
Description
ICAM3 Monoclonal antibody specifically detects ICAM3 in Human samples. It is validated for IHC-P, IF, Blocking, Flow Cyt.Specifications
ICAM3 | |
186-2G9 | |
intercellular adhesion molecule 3 | |
Liquid | |
Mouse | |
IgG2b, Kappa | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
Blocking Assay, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
ICAM3 | |
CD50, CDW50, ICAM-R | |
Stimulated human leukocytes | |
100 μg | |
Primary | |
3385 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title