Learn More
Selleck Chemical LLCSupplier Diversity Partner Icatibant Acetate DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S56952MG
Icatibant Acetate acts as an antagonist of the bradykinin B2 receptor and is used in the treatment of hereditary angioedema.
Catalog No. 50-194-8441