Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K1 Series Ice Cleat - Slim Profile

The K1 Slim is a one-size-fits-all traction solution designed for flat-soled footwear. The rotatable design combines a non-spiked anti-slip material with traditional ice-penetrating tungsten carbide studs to maximize safety and productivity when working indoors or outdoors.

$43.20

Specifications

Closure Type Elastic
Color Black
For Use With (Application) Outdoor Surfaces, Ice Surfaces
Size Universal
Strap Material Elastic
View More Specs

Products 2
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
19-060-836
View Documents
K1 Series
V9770650O/S
High Profile Tungston Carbide Molded in
Pair for $43.20
Only null left
Add to Cart
 
19-060-838
View Documents
K1 Series
V9770870O/S
Slim Profile Tungston Carbide Molded in
Pair for $43.20
Only null left
Add to Cart
 
Description

Description

  • Specially designed for flat-soled footwear found in walking and safety shoes
  • Rotatable design provides increased traction for both indoor and outdoor use
  • Non-spiked anti-slip material provides increased traction on wet surfaces
  • Ice-penetrating tungsten carbide studs maximize grip on ice and snow
  • Black or hi-viz adjustable strap delivers a perfect fit regardless of shoe size
  • Ultra-thin rubber pad provides maximum comfort and is designed for all-day use
Specifications

Specifications

Elastic
Outdoor Surfaces, Ice Surfaces
Elastic
Unisex
Black
Universal
Reusable
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.