Learn More
Cayman ChemicalSupplier Diversity Partner ICG-001, 780757-88-2, 10 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 1625710
A small molecule inhibitor of β-catenin/cyclic AMP responsive element binding protein binding protein (CBP)-mediated transcription (IC50 = 3 µM); selectively induces apoptosis in transformed colon cells but not in normal cells and prevents the growth of colon carcinoma cells at 25 µM in vitro
Catalog No. NC1463603