Learn More
Aat BioquestSupplier Diversity Partner ICG-ATT [3-ICG-acyl-1,3-thiazolidine-2-thione] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 181
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics. It is used for determining cardiac output, hepatic function, and liver blood flow, and for ophthalmic angiography. It has a peak spectral absorption close to 800 nm.
Catalog No. 50-195-2791