Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICI 182,780, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
ICI 182,780 is a high affinity estrogen receptor antagonist (IC50 = 0.29 nM), devoid of any partial agonism both in vitro and in vivo. ICI 182,780 is a high affinity agonist at the membrane estrogen receptor GPER. ICI 182,780 inhibits MCF-7 human breast cancer cell growth in vitro and shows antitumor activity in mice models. ICI 182,780 penetrates the blood-brain-barrier and has neuronal functional effects in vivo.
Specifications
Quantity | 1 mg |
Formula Weight | 606.77 |
Percent Purity | >99% |
Chemical Name or Material | ICI 182, 780 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction