Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ICI 204,448 hydrochloride
κ agonist, acts peripherally
$241.28 - $1008.04
Specifications
Chemical Name or Material | (RS)-[3-[1-[[(3,4-Dichlorophenyl)acetyl]methylamino]-2-(1-pyrrolidinyl)ethyl]phenoxy]acetic acid hydrochloride |
---|---|
CAS | 121264-04-8 |
Assay Percent Range | >98% |
Solubility | Soluble to 100mM in DMSO |
Molecular Formula | C23H26Cl2N2O4.HCl |
Description
Peripherally acting κ agonist.Specifications
(RS)-[3-[1-[[(3,4-Dichlorophenyl)acetyl]methylamino]-2-(1-pyrrolidinyl)ethyl]phenoxy]acetic acid hydrochloride | |
>98% | |
C23H26Cl2N2O4.HCl |
121264-04-8 | |
Soluble to 100mM in DMSO | |
Observed MW: 501.84 |
Safety and Handling
Recommended Storage : Store at Room Temperature
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title