Learn More
ICOS (CD278) Armenian Hamster, Alexa Fluor 647, Clone: C398.4A, BD Pharmingen™
Armenian Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 565883
Catalog No. BDB565883
Add to cart
Description
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
Specifications
ICOS (CD278)
C398.4A
Alexa Fluor 647
Purified
Armenian Hamster
Armenian Hamster IgG
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions, and unreacted Alexa Fluor™ 647 was removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115
Mouse D10.G4.1 T-cell clone
25μg
Cell Biology
Primary
Human, Murine, Rat, Rhesus Monkey
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.