Learn More
ICOS (CD278) Armenian Hamster, PE-CF594, Clone: C398.4A, BD Horizon™
Armenian Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 565889
 View more versions of this product
Catalog No. BDB565889
Add to cart
Description
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
Specifications
ICOS (CD278)
C398.4A
PE-CF594
Purified
Armenian Hamster
Armenian Hamster IgG
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115
Mouse D10.G4.1 T-cell clone
25μg
Cell Biology
Primary
Human, Murine, Rat, Rhesus Monkey
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.