Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS (CD278) Mouse anti-Human , Alexa Fluor 488, Clone: DX29, BD Biosciences™

Mouse Monoclonal Antibody

$94.10 - $232.50

Specifications

Antigen ICOS (CD278)
Clone DX29
Applications Flow Cytometry
Conjugate Alexa Fluor 488
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB567003
View Documents
BD Biosciences
567003
100 Tests
Each for $232.50
Add to cart
 
BDB567004
View Documents
BD Biosciences
567004
25 Tests
Each for $94.10
Add to cart
 
Description

Description

ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

ICOS (CD278)
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Activated human T cells
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed.
DX29
Alexa Fluor 488
Mouse
Human
ICOS; DX-29; H4; Inducible T-cell costimulator; AILIM; CVID1
Mouse IgG1, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit