Learn More
ICOS Mouse anti-Human, Alexa Fluor 700, Clone: 669222, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB6975N100UG
Catalog No. FAB6975N100
Add to cart
This item is not returnable. View return policy
Description
Description
ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ICOS
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q9Y6W8
ICOS
NS0 mouse myeloma cell line transfected with human ICOS and mouse myeloma cell line NS0-derived recombinant human ICOS cocktail Glu21-Phe141 Accession # Q9Y6W8
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human ICOS in direct ELISAs.
Flow Cytometry
669222
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Activation-inducible lymphocyte immunomediatory molecule, AILIM, AILIMCD278, CD278, CD278 antigen, CRP-1, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Mouse
IgG1
RUO
Primary
29851
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)