Learn More
ICOS Mouse anti-Human, Alexa Fluor 700, Clone: ISA-3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143126AF700
Catalog No. NBP143126AI
Add to cart
Description
Description
ICOS Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications
Specifications
ICOS
ISA-3
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ICOS
The immunogen for this antibody was ICOS.
Protein A or G purified
RUO
Primary
29851
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human