Learn More
ICOS Mouse anti-Human, DyLight 488, Clone: ISA-3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143126G
Catalog No. NBP143126G
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
ICOS Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications
Specifications
ICOS
ISA-3
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ICOS
The immunogen for this antibody was ICOS.
Protein A or G purified
RUO
Primary
29851
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human