Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS Mouse anti-Human, FITC, Clone: ISA-3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143126F

Catalog No. NBP143126F


Add to cart

Description

Description

ICOS Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications

Specifications

ICOS
ISA-3
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ICOS
The immunogen for this antibody was ICOS.
Protein A or G purified
RUO
Primary
29851
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
FITC
PBS with 0.05% Sodium Azide
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit