Alfa Aesar™ ICP-MS Memory Check Standard solution B for 6020 CLP-M, Specpure™

Manufacturer: Alfa Aesar™ 40488AP

Quantity 500mL

 View more versions of this product

Catalog No. AA40488AP

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00803323
    UN Number UN3264
    Chemical Name or Material ICP-MS Memory Check Standard solution B for 6020 CLP-M
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    MDL Number MFCD00803323
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Quantity 500mL
    TSCA TSCA
    Physical Form Liquid
    Concentration Matrix: 2% HNO3/tr. F-/tr. tartaric acid
    UN Number UN3264
    Health Hazard 1 Warning
    Density 1.0g/mL
    Chemical Name or Material ICP-MS Memory Check Standard solution B for 6020 CLP-M
    Grade Specpure™

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit