Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICT Antibody (1E9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242633AF350
Description
ICT Monoclonal specifically detects ICT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
ICT | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
MRPL58 | |
Recombinant human ICT1 (30-206aa) purified from E. coli | |
Cancer | |
3396 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA | |
1000000000 | |
Western Blot, Flow Cytometry, ELISA | |
Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial | |
Mouse | |
100 μL | |
Primary | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction