Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICT Mouse anti-Human, Alexa Fluor™ 750, Clone: 1000000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242633AF750

Catalog No. NP242633A70


Add to cart

Description

Description

ICT Monoclonal antibody specifically detects ICT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

ICT
1000000000
Western Blot, Flow Cytometry, ELISA
Conjugated, Purified
MRPL58
Recombinant human ICT1 (30-206aa) purified from E. coli
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial
Mouse
IgG1 Kappa
100 μL
Primary
3396.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit