Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICT1 Mouse anti-Human, Clone: OTI2F9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526220

Catalog No. PIMA526220


Add to cart

Description

Description

Essential peptidyl-tRNA hydrolase component of the mitochondrial large ribosomal subunit. Acts as a codon-independent translation release factor that has lost all stop codon specificity and directs the termination of translation in mitochondrion, possibly in case of abortive elongation. May be involved in the hydrolysis of peptidyl-tRNAs that have been prematurely terminated and thus in the recycling of stalled mitochondrial ribosomes.
Specifications

Specifications

ICT1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q14197
ICT1;DS1; Peptidyl-tRNA hydrolase ICT1, mitochondrial; Peptidyl-tRNA hydrolase ICT1, mitochondrial; Digestion substraction 1; Peptidyl-tRNA hydrolase ICT1, mitochondrial; Immature colon carcinoma transcript 1 protein
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3396
Immunohistochemistry (Paraffin), Western Blot
OTI2F9
Unconjugated
MRPL58
Liquid
MRPL58
Full length human recombit protein of ICT1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit