Learn More
Abbott™ ID NOW™ COVID-19 Control Kit
For use with ID NOW™ Instruments and Assay Kits.
Supplier:  Abbott™ 190080
Catalog No. 23-046-651
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • Includes 12 positive controls and 12 negative controls
  • For use with ID NOW™ Instruments and Assay Kits
Specifications
Specifications
COVID-19
ID NOW™ Instruments
Isothermal Molecular Technology
Yes
12 Positive Controls, 12 Negative Controls
RdRp
Kit
Nasal Swabs, Nasopharyngeal Swabs, Throat Swabs
Control
COVID Testing
24