Learn More
ID2 Mouse anti-Human, Alexa Fluor 488, Clone: 4E12G5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237417AF488
Catalog No. NBP237417Z
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ID2
4E12G5
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
ID2
Purified recombinant fragment of human ID2 (AA: 1-134) expressed in E. Coli.
Protein A or G purified
Cancer, Innate Immunity
Primary
3398
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human