Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ID2 Mouse anti-Human, Clone: A4-D4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266898
Description
ID2 Monoclonal antibody specifically detects ID2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ID2 | |
A4-D4 | |
Unconjugated | |
Purified | |
BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389 | |
ID2 | |
Recombinant protein | |
Monoclonal | |
Protein A purified | |
Cancer, Innate Immunity | |
Human,Mouse |
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin) | |
2 mg/mL | |
Western Blot 1:1000-1:10000 Immunohistochemistry 1:100 Immunocytochemistry/Immunofluorescence 1:100 Immunohistochemistry-Paraffin 1:100 | |
TBS (pH7.4), 0.5%BSA, 40% Glycerol with 0.05% Sodium Azide | |
3398 | |
Mouse | |
IgG2b | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title