Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID2 Mouse anti-Human, PE, Clone: 4E12G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237417PE

Catalog No. NBP237417PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ID2
4E12G5
Flow Cytometry
Purified
ID2
Purified recombinant fragment of human ID2 (AA: 1-134) expressed in E. Coli.
Protein A or G purified
Cancer, Innate Immunity
Primary
3398
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit