Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID3 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI8B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270979APCCY7

Catalog No. N270979APC7


Add to cart

Description

Description

ID3 Monoclonal antibody specifically detects ID3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ID3
OTI8B3
Flow Cytometry
Affinity Purified, Conjugated
ID3
Full length recombinant protein of human ID3 (NP_002158) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Cell Cycle and Replication, Prostate Cancer
Primary
3399.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
BHLHB25, bHLHb251R21, Class B basic helix-loop-helix protein 25, DNA-binding protein inhibitor ID-3, HEIR1, HEIR-1, Helix-loop-helix protein HEIR-1, ID-like protein inhibitor HLH 1R21, Inhibitor of DNA binding 3, inhibitor of DNA binding 3, dominant negative helix-loop-helix protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit