Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID3 Mouse anti-Human, Clone: OTI5C7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524998

Catalog No. PIMA524998


Add to cart

Description

Description

Members of the Id family of basic helix-loop-helix (bHLH) proteins include ID1, ID2, ID3 and ID4. They are ubiquitously expressed and dimerize with members of the class A and B HLH proteins. Due to the absence of the basic region, the resulting heterodimers cannot bind DNA. The ID-type proteins thus appear to negatively regulate DNA binding of bHLH proteins. Since ID1 inhibits DNA binding of E12 and Myo D, it apparently functions to inhibit muscle-specific gene expression. Under conditions that facilitate muscle cell differentiation, the ID protein levels fall, allowing E12 and/or E47 to form heterodimers with Myo D and myogenin, which in turn activate myogenic differentiation. It has been shown that expression of each of the ID proteins is strongly dependent on growth factor activation and that reduction of ID mRNA levels by antisense oligonucleotides leads to a delayed reentry of arrested cells into the cell cycle following growth factor stimulation.
Specifications

Specifications

ID3
Monoclonal
0.83 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q02535
1R21; bHLHb25; Class B basic helix-loop-helix protein 25; DNA binding protein inhibitor ID 3; DNA-binding protein inhibitor ID-3; HEIR 1; HEIR1; Helix loop helix protein HEIR 1; Helix loop helix protein HEIR1; Helix-loop-helix protein HEIR-1; HLH642; ID 3; ID like protein inhibitor HLH 1R21; ID like protein inhibitor HLH 462; ID-like protein inhibitor HLH 1R21; ID3; ID3_HUMAN; Inhibitor of DNA binding 3; Inhibitor of DNA binding 3 domit negative helix loop helix protein;
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3399
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI5C7
Unconjugated
ID3
Liquid
ID3
Full length human recombit protein of ID3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit