Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID4 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI2G10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270987AF700

Catalog No. N270987A700


Add to cart

Description

Description

ID4 Monoclonal antibody specifically detects ID4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ID4
OTI2G10
Western Blot
Affinity Purified, Conjugated
ID4
Full length recombinant protein of human ID4 (NP_001537) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein
Mouse
IgG1
0.1 mL
Primary
3400.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit