Learn More
Selleck Chemical LLCSupplier Diversity Partner Idarubicin HCl 10mg 57852-57-0 4-demethoxydaunorubicin (NSC256439, 4-DMDR) HCl  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S122810MG
Idarubicin HCl (4-demethoxydaunorubicin, NSC256439, 4-DMDR) is a hydrochloride salt form of Idarubicin which is an anthracycline antibiotic and a DNA topoisomerase II (topo II) inhibitor for MCF-7 cells with IC50 of 3.3 ng/mL in a cell-free assay. Idarubicin induces mTOR-dependent cytotoxic autophagy.
Catalog No. 50-651-0