Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDAS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549068

Catalog No. PA549068


Add to cart

Description

Description

Transcription regulator specifically required for multiciliate cell differentiation. Acts in a multiprotein complex containing E2F4 and E2F5 that binds and activates genes required for centriole biogenesis. Required for the deuterosome-mediated acentriolar pathway (PubMed:25048963). Plays a role in mitotic cell cycle progression by promoting cell cycle exit. Modulates GMNN activity by reducing its affinity for CDT1 (PubMed:21543332, PubMed:24064211).
Specifications

Specifications

IDAS
Polyclonal
PBS with 0.09% sodium azide
D6RGH6
Multicilin, Multiciliate differentiation and DNA synthesis-associated cell cycle protein, Protein Idas, MCIDAS, IDAS, MCI, MCIN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
345643
Western Blot
Unconjugated
MCIDAS
Liquid
MCIDAS
KLH conjugated synthetic peptide between 20-46 amino acids from the N-terminal region of human IDAS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit