Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idazoxan hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

79944-56-2
C11H13ClN2O2
240.687
MYUBYOVCLMEAOH-UHFFFAOYSA-N
2-(1, 4-Benzodioxan-2-yl)-2-imidazoline hydrochloride
154494

α2 antagonist. Also I2 ligand

α2-adrenoceptor antagonist, and I2 ligand, selective over I1 sites (pKi values are 5.90, 7.22, 8.01, 7.43, and 7.7 for I1, I2, α2A, α2B, and α2C receptors respectively).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

SMILES C1CN=C(N1)C2COC3=CC=CC=C3O2.Cl
IUPAC Name 2-(2,3-dihydro-1,4-benzodioxin-3-yl)-4,5-dihydro-1H-imidazole;hydrochloride
Formula Weight 240.69
Quantity 10mg
Chemical Name or Material Idazoxan hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature