Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idazoxan hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

79944-56-2
C11H13ClN2O2
240.687
MYUBYOVCLMEAOH-UHFFFAOYSA-N
2-(1, 4-Benzodioxan-2-yl)-2-imidazoline hydrochloride
154494

α2 antagonist. Also I2 ligand

α2-adrenoceptor antagonist, and I2 ligand, selective over I1 sites (pKi values are 5.90, 7.22, 8.01, 7.43, and 7.7 for I1, I2, α2A, α2B, and α2C receptors respectively).
SMILES C1CN=C(N1)C2COC3=CC=CC=C3O2.Cl
IUPAC Name 2-(2,3-dihydro-1,4-benzodioxin-3-yl)-4,5-dihydro-1H-imidazole;hydrochloride
Formula Weight 240.69
Quantity 50mg
Chemical Name or Material Idazoxan hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature