Learn More
BIOGEMS INTERNATIONAL INC IDE1 1mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 11648991MG
Encompass_Preferred
IDE1 activates the TGF-beta signaling pathway and induces definitive endoderm formation in human and mouse embryonic stem cells. It is also reported to increase the levels of Nodal expression, induce Smad2 phosphorylation and SOX17 expression.
Catalog No. 50-152-9666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.