Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idebenone, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
58186-27-9
C19H30O5
338.444
JGPMMRGNQUBGND-UHFFFAOYSA-N
idebenone,idebenona,idebenonum,idebenonum latin,idebenona spanish,idebenone inn:jan,unii-hb6pn45w4j,2-10-hydroxydecyl-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione,6-10-hydroxydecyl-2,3-dimethoxy-5-methyl-1,4-benzoquinone,hb6pn45w4j
3686
CHEBI:31687
2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
CC1=C(C(=O)C(=C(C1=O)OC)OC)CCCCCCCCCCO
This item is not returnable. View return policy
58186-27-9
C19H30O5
338.444
JGPMMRGNQUBGND-UHFFFAOYSA-N
idebenone,idebenona,idebenonum,idebenonum latin,idebenona spanish,idebenone inn:jan,unii-hb6pn45w4j,2-10-hydroxydecyl-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione,6-10-hydroxydecyl-2,3-dimethoxy-5-methyl-1,4-benzoquinone,hb6pn45w4j
3686
CHEBI:31687
2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
CC1=C(C(=O)C(=C(C1=O)OC)OC)CCCCCCCCCCO

Antioxidant and neuroprotective agent

Antioxidant and neuroprotective agent. Protects mitochondrial membranes against lipid peroxidation and blocks glutamate neurotoxicity in vitro and in vivo. Inhibits apoptosis of astrocytes via increased NGF production.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 338.44
Percent Purity >99%
Chemical Name or Material Idebenone

Recommended Storage : Store at Room Temperature