Learn More
Idex Stainless-Steel Tubing
Encompass
Precut stainless steel tubing ready for all chromatography testing requirements
$36.18
Specifications
Autoclavable Autoclavable
Certifications/Compliance ASTM A269 and A213
View More Specs
Products 1
Catalog Number Mfr. No. Overall Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Overall Length (Metric) Price Quantity & Availability  
05-701-139
Encompass
View Documents Promotion Details
Idex
U148
30 cm
Each for $36.18
Add to cart
Description
Description
  • 316 stainless steel tubing, polished and passivated
  • Color coded for easy identification
  • Suited for most analytical applications
  • Max pressure 5800 psi
Specifications
Specifications
Autoclavable
ASTM A269 and A213