Learn More
Idex Syringe Sample Filters
Designed specifically to remove particles from analytical HPLC samples
Manufacturer:  Idex B100
 View more versions of this product
Catalog No. 05-707-61
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Polypropylene holder incorporates 0.79mm (0.03 in.) thick, 3mm (.12 in.) diameter stainless steel frit, creating very little back pressure
  • Excellent system protection
  • Ultra-low hold-up volume
  • Disposable
Specifications
Specifications
2μm
Syringe Sample Filter