Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDH1 Monoclonal Antibody (CL0219)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA534971
Description
Immunogen sequence: FVVPGPGKVE ITYTPSDGTQ KVTYLVHNFE EGGGVAMGMY NQDKSIEDFA HSSFQMALSK GWPLY.
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the cytoplasm and peroxisomes. It contains the PTS-1 peroxisomal targeting signal sequence. The presence of this enzyme in peroxisomes suggests roles in the regeneration of NADPH for intraperoxisomal reductions, such as the conversion of 2, 4-dienoyl-CoAs to 3-enoyl-CoAs, as well as in peroxisomal reactions that consume 2-oxoglutarate, namely the alpha-hydroxylation of phytanic acid. The cytoplasmic enzyme serves a significant role in cytoplasmic NADPH production.
Specifications
IDH1 | |
Monoclonal | |
Unconjugated | |
IDH1 | |
AI314845; AI788952; cb876; corneal epithelial crystallin; cytosolic NADP+-dependent isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrongenase; E030024J03Rik; epididymis luminal protein 216; epididymis secretory protein Li 26; fm90e09; HEL-216; HEL-S-26; ICDH; Id-1; IDCD; IDH; Idh1; Idh-1; IDP; Idpc; im:7143416; isocitrate dehydrogenase (NAD(+)) IDH1; isocitrate dehydrogenase (NADP(+)) 1; isocitrate dehydrogenase (NADP(+)) 1, cytosolic; Isocitrate dehydrogenase [NAD] subunit 1, mitochondrial; Isocitrate dehydrogenase [NADP] cytoplasmic; isocitrate dehydrogenase 1 (NADP+); isocitrate dehydrogenase 1 (NADP+), soluble; Isocitrate dehydrogenase 1, soluble; Isocitric dehydrogenase; N2690; NAD(+)-specific ICDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP-cICDH; NADP-dependent isocitrate dehydrogenase, cytosolic; NADP-dependent isocitrate dehydrogenase, peroxisomal; OTTHUMP00000206464; OTTHUMP00000206465; OTTHUMP00000206466; oxalosuccinate decarboxylase; PICD; wu:fm90e09; YNL037C | |
Mouse | |
Protein A | |
RUO | |
3417 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
CL0219 | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75874 | |
IDH1 | |
Recombinant Protein corresonding to Human IDH1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction